Assay Details
| Gene Symbol: Adamdec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021475.2 | NP_067450.1 | 12 - 13 | 1338 | 97 | |
| GenBank mRNA | AJ242912.1 | - | 12 - 13 | 1338 | 97 | |
| AK008929.1 | - | 3 - 4 | 349 | 97 | ||
| AK156586.1 | - | 12 - 13 | 1353 | 97 | ||
| BC026542.1 | - | 12 - 13 | 1362 | 97 | ||
| BC046324.1 | - | 12 - 13 | 1329 | 6394673 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamdec1
Genomic Map