Assay Details
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026314.3 | NP_080590.3 | 4 - 5 | 727 | 131 | |
GenBank mRNA | AK005832.1 | - | 1 - 2 | 177 | 131 | |
AY428507.1 | - | 4 - 5 | 727 | 131 | ||
BC026462.1 | - | 4 - 5 | 684 | 4483015 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
