Assay Details
| Gene Symbol: Uckl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026765.3 | NP_081041.2 | 12 - 13 | 1312 | 82 | |
| GenBank mRNA | AK003541.1 | - | 2 - 3 | 158 | 82 | |
| BC016535.1 | - | 12 - 13 | 1312 | 2655873 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uckl1
Genomic Map