Assay Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027315.4 | NP_081591.3 | 2 - 3 | 535 | 133 | |
| GenBank mRNA | AK009324.1 | - | 1 - 2 | 117 | 133 | |
| AK146707.1 | - | 2 - 3 | 348 | 133 | ||
| AK214106.1 | - | 1 - 2 | 56 | 133 | ||
| AY945937.1 | - | 2 - 3 | 432 | 133 | ||
| BC082275.1 | - | 2 - 3 | 358 | 6310111 | 133 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC082275.1 transcripts.
Set Membership: |
|

Ube2q1
Genomic Map