Assay Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027315.4 | NP_081591.3 | 4 - 5 | 693 | 97 | |
| GenBank mRNA | AK009324.1 | - | 3 - 4 | 275 | 97 | |
| AK146707.1 | - | 4 - 5 | 506 | 97 | ||
| AK214106.1 | - | 3 - 4 | 214 | 97 | ||
| AY112699.1 | - | 4 - 5 | 686 | 97 | ||
| AY945937.1 | - | 4 - 5 | 590 | 97 | ||
| BC060966.1 | - | 2 - 3 | 86 | 97 | ||
| BC082275.1 | - | 4 - 5 | 516 | 6310111 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2q1
Genomic Map