Assay Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027315.4 | NP_081591.3 | 5 - 6 | 832 | 101 | |
| GenBank mRNA | AK009324.1 | - | 4 - 5 | 414 | 101 | |
| AK146707.1 | - | 5 - 6 | 645 | 101 | ||
| AY112699.1 | - | 5 - 6 | 825 | 101 | ||
| AY945937.1 | - | 5 - 6 | 729 | 101 | ||
| BC030044.1 | - | 2 - 3 | 157 | 101 | ||
| BC060966.1 | - | 3 - 4 | 225 | 101 | ||
| BC082275.1 | - | 5 - 6 | 655 | 6310111 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2q1
Genomic Map