Assay Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027315.4 | NP_081591.3 | 7 - 8 | 984 | 110 | |
| GenBank mRNA | AK009324.1 | - | 6 - 7 | 566 | 110 | |
| AK146707.1 | - | 7 - 8 | 797 | 110 | ||
| AY112699.1 | - | 7 - 8 | 977 | 110 | ||
| AY945937.1 | - | 7 - 8 | 881 | 110 | ||
| BC030044.1 | - | 4 - 5 | 309 | 110 | ||
| BC060966.1 | - | 5 - 6 | 377 | 110 | ||
| BC082275.1 | - | 7 - 8 | 807 | 6310111 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2q1
Genomic Map