Assay Details
| Gene Symbol: Stk11ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027886.3 | NP_082162.3 | 7 - 8 | 758 | 78 | |
| GenBank mRNA | AK004757.2 | - | 7 - 8 | 711 | 78 | |
| AK156122.1 | - | 6 - 7 | 846 | 78 | ||
| AK165911.1 | - | 7 - 8 | 749 | 78 | ||
| AK171989.1 | - | 6 - 7 | 620 | 78 | ||
| BC042403.1 | - | 6 - 7 | 617 | 78 | ||
| BC061635.1 | - | 7 - 8 | 6530178 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk11ip
Genomic Map