Assay Details
| Gene Symbol: Stk11ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027886.3 | NP_082162.3 | 18 - 19 | 2424 | 68 | |
| GenBank mRNA | AK004757.2 | - | 18 - 19 | 2377 | 68 | |
| AK156122.1 | - | 17 - 18 | 2512 | 68 | ||
| AK165911.1 | - | 18 - 19 | 2416 | 68 | ||
| AK171989.1 | - | 17 - 18 | 2286 | 68 | ||
| AK213443.1 | - | 2 - 3 | 330 | 68 | ||
| BC035530.1 | - | 4 - 5 | 1464 | 68 | ||
| BC042403.1 | - | 16 - 17 | 2461 | 68 | ||
| BC061635.1 | - | 18 - 19 | 2407 | 6530178 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk11ip
Genomic Map