Assay Details
| Gene Symbol: Dhdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027903.3 | NP_082179.1 | 3 - 4 | 397 | 73 | |
| GenBank mRNA | AK005050.2 | - | 3 - 4 | 398 | 73 | |
| AK154452.1 | - | 3 - 4 | 379 | 73 | ||
| BC031710.1 | - | 3 - 4 | 375 | 73 | ||
More Information
Set Membership: |
|

Dhdh
Genomic Map