Assay Details
Gene Symbol: Dhdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027903.3 | NP_082179.1 | 6 - 7 | 922 | 101 | |
GenBank mRNA | AK005050.2 | - | 6 - 7 | 923 | 101 | |
AK154452.1 | - | 6 - 7 | 904 | 101 | ||
BC031710.1 | - | 6 - 7 | 900 | 101 | ||
BC116414.2 | - | 6 - 7 | 906 | 40103176 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25093406 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25093407 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25093408 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039071 | BC031710.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039071 | AK154452.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039071 | NM_027903.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039071 | AK005050.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560935 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697510 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780267 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880438 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675875 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955200 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61409 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161611 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170582 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039071 | BC031710.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039071 | AK154452.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039071 | NM_027903.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039071 | AK005050.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P365351 | BC031710.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P365351 | AK154452.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P365351 | NM_027903.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P365351 | AK005050.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093413 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093421 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093409 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093398 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093396 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093407 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093395 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093405 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25093428 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06183095 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01507839 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170582 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161611 | - | - |
Whole Mouse Genome G4122A | A_51_P365351 | BC031710.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P365351 | AK154452.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P365351 | NM_027903.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P365351 | AK005050.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P220743 | AK032449.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161611 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170582 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06529751 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06108016 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25093408 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25093407 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25093406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25093370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06324605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06495414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06440429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06238669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06324604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06303938 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06420779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06351143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06108016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093354 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06121508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05892111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25093386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06351151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06186552 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10563321 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563322 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563316 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563320 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563318 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563319 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563317 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563315 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453487_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10563314 | - | - |
Mouse Genome 430 2.0 Array | 1453487_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453487_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71755_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71755_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700027249.mm.1 | NM_027903.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700027249.mm.1 | NM_027903.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700027249.mm.1 | TC0700003008.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700027250.mm.1 | NM_027903.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700027250.mm.1 | NM_027903.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700027250.mm.1 | TC0700003008.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700027251.mm.1 | NM_027903.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700027251.mm.1 | NM_027903.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700027251.mm.1 | TC0700003008.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453487_PM_at | - | - |
Mouse Expression Set 430 | 1453487_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453487_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453487_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 759431 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 870193 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6940471 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6840520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 870193 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006940471 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000870193 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006840520 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241717 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234086 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689940 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2689940 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1234086 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1241717 | - | - |
mouse-6 v1.1 (Illumina) | scl00033.1_17-S | - | - |
mouse-6 v1.1 (Illumina) | scl071755.1_49-S | - | - |
mouse-6 v1.1 (Illumina) | scl0071755.2_316-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 870193 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6940471 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6840520 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0071755.2_316-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl071755.1_49-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00033.1_17-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940471 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840520 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 870193 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2971314 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1234086 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2689940 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2689939 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2971314 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689940 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234086 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689939 | - | - |
