Assay Details
| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080995.1 | NP_001074464.1 | 4 - 5 | 377 | 75 | |
| GenBank mRNA | AK014608.2 | - | 3 - 4 | 297 | 75 | |
| AK154915.1 | - | 3 - 4 | 280 | 75 | ||
| BC066148.1 | - | 3 - 4 | 271 | 30086532 | 75 | |
| DQ400346.1 | - | 3 - 4 | 311 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
cellular response to DNA damage stimulus
cell cycle
cell cycle arrest
spermatid development
hemopoiesis
cellular response to UV
cellular response to cytokine stimulus
cellular response to hydroperoxide
cellular response to gamma radiation
mitotic cell cycle arrest
negative regulation of fibroblast apoptotic process
molecular_function
apoptotic process
cellular response to DNA damage stimulus
cell cycle
cell cycle arrest
spermatid development
hemopoiesis
cellular response to UV
cellular response to cytokine stimulus
cellular response to hydroperoxide
cellular response to gamma radiation
mitotic cell cycle arrest
negative regulation of fibroblast apoptotic process
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369252 | AK014608.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369252 | AK014608.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773804 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144767 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168493 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155389 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369252 | AK014608.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369252 | AK014608.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168493 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06282435 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155389 | - | - |
| Whole Mouse Genome G4122A | A_51_P369252 | AK014608.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369252 | AK014608.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111212 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155389 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168493 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06031190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06031178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06031191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06031193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06031171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25326970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25327163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06056589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06150896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06031170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06175484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06266007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06567663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06128684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06031169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06031188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25326974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25327024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10565572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565575 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565571 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434240_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429734_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10565570 | - | - |
| Mouse Genome 430 2.0 Array | 1434240_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429734_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429734_at | - | - |
| Mouse Genome 430 2.0 Array | 1434240_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74041_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74041_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700031557.mm.1 | NM_001080995.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700031557.mm.1 | TC0700003743.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434240_PM_at | - | - |
| Mouse Expression Set 430 | 1429734_at | - | - |
| Mouse Expression Set 430 | 1429734_PM_at | - | - |
| Mouse Expression Set 430 | 1434240_at | - | - |
| Murine Genome U74v2 Set | 132418_at | - | - |
| Murine Genome U74v2 Set | 132418_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434240_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429734_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429734_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434240_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 817048 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2230451 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2660829 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2446902 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2660829 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2660829 | - | - |

Ddias
Genomic Map