Assay Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195025.1 | NP_001181954.1 | 5 - 6 | 706 | 89 | |
| NM_028778.4 | NP_083054.1 | 4 - 5 | 682 | 89 | ||
| GenBank mRNA | AK004737.2 | - | 4 - 5 | 670 | 89 | |
| AK033672.1 | - | 4 - 5 | 669 | 89 | ||
| AK034082.1 | - | 5 - 6 | 698 | 89 | ||
| BC033302.1 | - | 4 - 5 | 657 | 3663595 | 89 | |
| BC046833.1 | - | 5 - 6 | 681 | 6816569 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nuak2
Genomic Map