Assay Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195025.1 | NP_001181954.1 | 6 - 7 | 836 | 60 | |
| NM_028778.4 | NP_083054.1 | 5 - 6 | 812 | 60 | ||
| GenBank mRNA | AK004737.2 | - | 5 - 6 | 800 | 60 | |
| AK033672.1 | - | 5 - 6 | 799 | 60 | ||
| AK034082.1 | - | 6 - 7 | 828 | 60 | ||
| BC033302.1 | - | 5 - 6 | 787 | 3663595 | 60 | |
| BC046833.1 | - | 6 - 7 | 811 | 6816569 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nuak2
Genomic Map