Assay Details
| Gene Symbol: Acbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102436.1 | NP_001095906.1 | 11 - 12 | 1474 | 68 | |
| NM_001102437.1 | NP_001095907.1 | 11 - 12 | 1604 | 68 | ||
| NM_001102438.1 | NP_001095908.1 | 11 - 12 | 1601 | 68 | ||
| NM_028793.3 | NP_083069.1 | 11 - 12 | 1477 | 68 | ||
| GenBank mRNA | AK005001.2 | - | 10 - 11 | 1393 | 68 | |
| AK050450.1 | - | 11 - 12 | 1440 | 68 | ||
| AK147839.1 | - | 11 - 12 | 1604 | 68 | ||
| AK214423.1 | - | 2 - 3 | 158 | 68 | ||
| BC035202.1 | - | 11 - 12 | 1530 | 68 | ||
| BC053518.1 | - | 11 - 12 | 1568 | 68 | ||
| BC061484.1 | - | 10 - 11 | 1375 | 5355466 | 68 | |
| BC084584.1 | - | 11 - 12 | 1406 | 5257773 | 68 | |
| BC090641.1 | - | 11 - 12 | 1428 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acbd5
Genomic Map