Assay Details
| Gene Symbol: P2ry13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028808.3 | NP_083084.2 | 1 - 2 | 67 | 81 | |
| GenBank mRNA | AK008013.2 | - | 1 - 2 | 68 | 81 | |
| AK038560.1 | - | 1 - 2 | 68 | 81 | ||
| AK083264.1 | - | 1 - 2 | 68 | 81 | ||
| BC119043.1 | - | 1 - 2 | 64 | 8733792 | 81 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008013.2,AK038560.1,AK083264.1,BC119043.1,NM_028808.3 transcripts.
Set Membership: |
|

P2ry13
Genomic Map