Assay Details
| Gene Symbol: Arhgap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039692.1 | NP_001034781.1 | 16 - 17 | 2516 | 93 | |
| NM_029277.2 | NP_083553.2 | 15 - 16 | 2441 | 93 | ||
| GenBank mRNA | AK031648.1 | - | 16 - 17 | 2360 | 93 | |
| AK077063.1 | - | 16 - 17 | 2517 | 93 | ||
| BC024633.1 | - | 11 - 12 | 928 | 93 | ||
| BC037608.1 | - | 15 - 16 | 2256 | 93 | ||
| BC141184.1 | - | 15 - 16 | 2250 | 9055796 | 93 | |
| BC145506.1 | - | 15 - 16 | 2249 | 9054069 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap12
Genomic Map