Assay Details
| Gene Symbol: Smurf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038627.1 | NP_001033716.1 | 8 - 9 | 1076 | 69 | |
| NM_029438.3 | NP_083714.3 | 8 - 9 | 1076 | 69 | ||
| GenBank mRNA | AK015264.1 | - | 4 - 5 | 475 | 69 | |
| CB524571.1 | - | 2 - 3 | 154 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smurf1
Genomic Map