Assay Details
| Gene Symbol: Mrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029844.3 | NP_084120.1 | 1 - 2 | 344 | 59 | |
| GenBank mRNA | AK003912.2 | - | 1 - 2 | 345 | 59 | |
| AK158113.1 | - | 1 - 2 | 218 | 59 | ||
| AY079153.1 | - | 1 - 2 | 121 | 59 | ||
| BC027543.1 | - | 1 - 2 | 198 | 1477097 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cAMP biosynthetic process
protein localization to cell surface
brown fat cell differentiation
corticotropin hormone receptor binding
type 3 melanocortin receptor binding
type 4 melanocortin receptor binding
type 5 melanocortin receptor binding
type 1 melanocortin receptor binding
positive regulation of cAMP biosynthetic process
protein localization to cell surface
brown fat cell differentiation
corticotropin hormone receptor binding
type 3 melanocortin receptor binding
type 4 melanocortin receptor binding
type 5 melanocortin receptor binding
type 1 melanocortin receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | AK158113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | AY079153.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | AK003912.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | BC027543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | NM_029844.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | AK158113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | AY079153.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | AK003912.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | BC027543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P346964 | NM_029844.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 138859 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162400 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116316 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | AK158113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | AY079153.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | AK003912.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | BC027543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | NM_029844.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | AK158113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | AY079153.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | AK003912.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | BC027543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P346964 | NM_029844.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30958586 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162400 | - | - |
| Whole Mouse Genome G4122A | A_51_P346964 | AK158113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | AY079153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | AK003912.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | BC027543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | NM_029844.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | AK158113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | AY079153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | AK003912.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | BC027543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346964 | NM_029844.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112550 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116316 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162400 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12677554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12677561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12677548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12677547 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12677550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30958727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12775546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12677550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12619610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12861786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12868876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12855084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13013546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12727282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12775549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12677547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12677554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12619606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12726138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30958579 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10436808 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436807 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436805 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436806 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451371_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451966_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10436804 | - | - |
| Mouse Genome 430 2.0 Array | 1451371_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451371_at | - | - |
| Mouse Genome 430 2.0 Array | 1451966_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451966_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77037_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77037_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600007651.mm.1 | NM_029844.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007651.mm.1 | NM_029844.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007651.mm.1 | TC1600000988.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600007650.mm.1 | NM_029844.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007650.mm.1 | NM_029844.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007650.mm.1 | TC1600000988.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600007655.mm.1 | NM_029844.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007655.mm.1 | NM_029844.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007655.mm.1 | TC1600000988.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600007657.mm.1 | NM_029844.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007657.mm.1 | NM_029844.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007657.mm.1 | TC1600000988.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600007656.mm.1 | NM_029844.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007656.mm.1 | NM_029844.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600007656.mm.1 | TC1600000988.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451966_at | - | - |
| Mouse Expression Set 430 | 1451966_PM_at | - | - |
| Mouse Expression Set 430 | 1451371_PM_at | - | - |
| Mouse Expression Set 430 | 1451371_at | - | - |
| Murine Genome U74v2 Set | 104325_PM_at | - | - |
| Murine Genome U74v2 Set | 104325_at | - | - |
| Murine Genome U74v2 Set | 171065_at | - | - |
| Murine Genome U74v2 Set | 171065_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451966_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451371_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451371_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451966_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451371_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451966_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451371_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451966_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 752295 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2940114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2940114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940114 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234241 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234241 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001829.1_310-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001829.1_310-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234241 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234241 | - | - |

Mrap
Genomic Map