Assay Details
| Gene Symbol: Crispld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030209.3 | NP_084485.1 | 10 - 11 | 1313 | 84 | |
| GenBank mRNA | AK019034.1 | - | 10 - 11 | 1309 | 84 | |
| AK033858.1 | - | 10 - 11 | 1277 | 84 | ||
| AK132171.1 | - | 10 - 11 | 1256 | 84 | ||
| AK149909.1 | - | 10 - 11 | 1270 | 84 | ||
| AK149925.1 | - | 10 - 11 | 1270 | 84 | ||
| AK150180.1 | - | 10 - 11 | 1270 | 84 | ||
| AK154308.1 | - | 10 - 11 | 1313 | 84 | ||
| BC090642.1 | - | 10 - 11 | 1307 | 30653388 | 84 | |
More Information
Set Membership: |
|

Crispld2
Genomic Map