Assay Details
| Gene Symbol: Rpp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133982.1 | NP_598743.1 | 1 | 1216 | 120 | |
| GenBank mRNA | AK082822.1 | - | 1 | 1287 | 120 | |
| AK140177.1 | - | 1 | 1293 | 120 | ||
| BC016085.1 | - | 1 | 1216 | 4501535 | 120 | |
| BC083064.1 | - | 1 | 1276 | 5367203 | 120 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542153 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542157 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542150 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542152 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542155 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157186 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542121 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157186 | - | - |
| Whole Mouse Genome G4122A | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104392 | AK140177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104392 | BC016085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104392 | BC083064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104392 | NM_133982.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111674 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103685 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07748500 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07748497 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07748510 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07748493 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07748509 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07748496 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542155 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542152 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542153 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542157 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542150 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07748475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07748499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07601661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07748510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08033728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07533758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07748465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07748515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07748462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07988083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07872189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542167 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10585704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585705 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448845_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10585703 | - | - |
| Mouse Genome 430 2.0 Array | 1448845_at | - | - |
| Mouse Genome 430 2.0 Array | 1448845_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102614_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102614_PM_at | - | - |
| Mouse Expression Set 430 | 1448845_at | - | - |
| Mouse Expression Set 430 | 1448845_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448845_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448845_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448845_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448845_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 738217 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6620048 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6620048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2754222 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2754222 | - | - |
| mouse-6 v1.1 (Illumina) | scl36905.1.71_309-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36905.1.71_309-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2754222 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2754222 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36905.1.71_309-S | - | - |

Rpp25
Genomic Map