Assay Details
| Gene Symbol: Mfsd6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146004.1 | NP_666116.1 | 1 | 52 | 66 | |
| GenBank mRNA | BC024997.1 | - | 1 | 52 | 5008948 | 66 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC024997.1,NM_146004.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27890038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27890039 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P475938 | NM_146004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P475938 | BC024997.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P475938 | NM_146004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P475938 | BC024997.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469401 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114956 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P475938 | NM_146004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P475938 | BC024997.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P475938 | NM_146004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P475938 | BC024997.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27890037 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114956 | - | - |
| Whole Mouse Genome G4122A | A_51_P475938 | NM_146004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475938 | BC024997.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475938 | NM_146004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475938 | BC024997.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127101 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09359846 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09359852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09359875 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09359845 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09359854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27890038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27890039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27890087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09359848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09588249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09439040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09359856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09359859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09479859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09233381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09359873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09359872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09594116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09588257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09231160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27890085 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10377309 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425302_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377308 | - | - |
| Mouse Genome 430 2.0 Array | 1425302_at | - | - |
| Mouse Genome 430 2.0 Array | 1425302_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215723_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215723_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100007287.mm.1 | NM_146004.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100007287.mm.1 | NM_146004.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100007287.mm.1 | TC1100000874.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425302_at | - | - |
| Mouse Expression Set 430 | 1425302_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425302_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425302_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425302_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425302_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 677919 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 610711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000610711 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247389 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247389 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2525942 | - | - |
| mouse-6 v1.1 (Illumina) | scl41391.1.1_282-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 610711 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41391.1.1_282-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610711 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247389 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247389 | - | - |

Mfsd6l
Genomic Map