Assay Details
| Gene Symbol: Dner | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152915.1 | NP_690879.1 | 1 - 2 | 769 | 86 | |
| GenBank mRNA | AB067650.1 | - | 1 - 2 | 418 | 86 | |
| AF370126.1 | - | 1 - 2 | 425 | 86 | ||
| AK030642.1 | - | 1 - 2 | 568 | 86 | ||
| AK044597.1 | - | 1 - 2 | 532 | 86 | ||
| AY032924.1 | - | 1 - 2 | 425 | 86 | ||
| BC034634.1 | - | 1 - 2 | 769 | 5365190 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dner
Genomic Map