Assay Details
| Gene Symbol: Dner | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152915.1 | NP_690879.1 | 11 - 12 | 2349 | 53 | |
| GenBank mRNA | AB067650.1 | - | 11 - 12 | 1998 | 53 | |
| AF370126.1 | - | 11 - 12 | 2005 | 53 | ||
| AK044597.1 | - | 11 - 12 | 2112 | 53 | ||
| AK152755.1 | - | 6 - 7 | 752 | 53 | ||
| AK153235.1 | - | 6 - 7 | 752 | 53 | ||
| AY032924.1 | - | 11 - 12 | 2005 | 53 | ||
| BC022636.1 | - | 6 - 7 | 812 | 53 | ||
| BC034634.1 | - | 11 - 12 | 2349 | 5365190 | 53 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dner
Genomic Map