| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500012370.mm.1 |
NM_152923.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012370.mm.1 |
NM_152923.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012370.mm.1 |
TC1500001646.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012368.mm.1 |
NM_152923.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012368.mm.1 |
NM_152923.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012368.mm.1 |
TC1500001646.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012369.mm.1 |
NM_152923.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012369.mm.1 |
NM_152923.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012369.mm.1 |
TC1500001646.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012374.mm.1 |
NM_152923.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012374.mm.1 |
NM_152923.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012374.mm.1 |
TC1500001646.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012373.mm.1 |
NM_152923.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012373.mm.1 |
NM_152923.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012373.mm.1 |
TC1500001646.mm.1 |
MTA TC |