Assay Details
Gene Symbol: Rnf113a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153503.1 | NP_705723.1 | 1 | 56 | 67 | |
GenBank mRNA | AK146283.1 | - | 1 | 155 | 67 | |
BC024906.1 | - | 1 | 56 | 3495576 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK146283.1,BC024906.1,NM_153503.1 transcripts.
This assay is in the 5'UTR region of the AK146283.1,BC024906.1,NM_153503.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322126 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322125 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322123 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322124 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | AK013178.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | AK146283.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | BC024906.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | AK013178.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | AK146283.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285487 | BC024906.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55054 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120351 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | AK013178.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | AK146283.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | BC024906.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | AK013178.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | AK146283.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285487 | BC024906.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | AK013178.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | AK146283.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | BC024906.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | AK013178.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | AK146283.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285487 | BC024906.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322142 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322132 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322145 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322143 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32322139 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120351 | - | - |
Whole Mouse Genome G4122A | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285487 | AK013178.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285487 | AK146283.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285487 | BC024906.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285487 | NM_153503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285487 | AK013178.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285487 | AK146283.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285487 | BC024906.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127194 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120351 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14555194 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14555208 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14555205 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322126 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322123 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322124 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32322125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14918045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14858431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14644096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322133 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15199717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322122 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322139 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10599245 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424837_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10599243 | - | - |
Mouse Genome 430 2.0 Array | 1424837_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424837_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69942_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69942_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00002236.mm.1 | NM_153503.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00002236.mm.1 | TC0X00000337.mm.1 | MTA TC |
Mouse Expression Set 430 | 1424837_PM_at | - | - |
Mouse Expression Set 430 | 1424837_at | - | - |
Murine Genome U74v2 Set | 170330_PM_at | - | - |
Murine Genome U74v2 Set | 167363_PM_f_at | - | - |
Murine Genome U74v2 Set | 170330_at | - | - |
Murine Genome U74v2 Set | 167363_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1424837_at | - | - |
Mouse Genome 430A 2.0 Array | 1424837_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424837_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424837_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 841683 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2650520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2650520 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002650520 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237083 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237083 | - | - |
mouse-6 v1.1 (Illumina) | scl54988.1.64_253-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2650520 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl54988.1.64_253-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650520 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2588728 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2588728 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54988.1.64_253-S | - | - |
