Assay Details
| Gene Symbol: Adap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172133.1 | NP_742145.1 | 6 - 7 | 717 | 72 | |
| GenBank mRNA | AK050079.1 | - | 6 - 7 | 775 | 72 | |
| AK050398.1 | - | 6 - 7 | 766 | 72 | ||
| AK053152.1 | - | 6 - 7 | 761 | 72 | ||
| AK139005.1 | - | 6 - 7 | 769 | 72 | ||
| AK140864.1 | - | 6 - 7 | 748 | 72 | ||
| BC027165.1 | - | 6 - 7 | 717 | 4951680 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adap2
Genomic Map