Assay Details
| Gene Symbol: Ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172146.2 | NP_742158.1 | 2 - 3 | 690 | 107 | |
| GenBank mRNA | AK147750.1 | - | 2 - 3 | 340 | 107 | |
| AK148034.1 | - | 2 - 3 | 294 | 107 | ||
| AK159141.1 | - | 3 - 4 | 840 | 107 | ||
| AK159859.1 | - | 3 - 4 | 840 | 107 | ||
| AK160542.1 | - | 2 - 3 | 315 | 107 | ||
| AK164506.1 | - | 2 - 3 | 304 | 107 | ||
| AK167057.1 | - | 2 - 3 | 329 | 107 | ||
| BC006053.1 | - | 2 - 3 | 277 | 107 | ||
| BC023841.1 | - | 2 - 3 | 317 | 5346105 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppat
Genomic Map