Assay Details
| Gene Symbol: Ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172146.2 | NP_742158.1 | 3 - 4 | 897 | 111 | |
| GenBank mRNA | AK147750.1 | - | 3 - 4 | 547 | 111 | |
| AK148034.1 | - | 3 - 4 | 501 | 111 | ||
| AK159141.1 | - | 4 - 5 | 1047 | 111 | ||
| AK159859.1 | - | 4 - 5 | 1047 | 111 | ||
| AK160542.1 | - | 3 - 4 | 522 | 111 | ||
| AK164506.1 | - | 3 - 4 | 511 | 111 | ||
| AK167057.1 | - | 3 - 4 | 536 | 111 | ||
| BC023841.1 | - | 3 - 4 | 524 | 5346105 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppat
Genomic Map