Assay Details
| Gene Symbol: Ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172146.2 | NP_742158.1 | 9 - 10 | 1729 | 139 | |
| GenBank mRNA | AK147750.1 | - | 9 - 10 | 1379 | 139 | |
| AK148034.1 | - | 9 - 10 | 1333 | 139 | ||
| AK159141.1 | - | 10 - 11 | 1879 | 139 | ||
| AK159859.1 | - | 10 - 11 | 1879 | 139 | ||
| AK160542.1 | - | 9 - 10 | 1354 | 139 | ||
| AK164506.1 | - | 9 - 10 | 1343 | 139 | ||
| AK167057.1 | - | 9 - 10 | 1368 | 139 | ||
| BC006053.1 | - | 8 - 9 | 1571 | 139 | ||
| BC023841.1 | - | 9 - 10 | 1356 | 5346105 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppat
Genomic Map