Assay Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289668.1 | NP_001276597.1 | 18 - 19 | 2022 | 70 | |
| NM_172257.4 | NP_758461.1 | 17 - 18 | 1959 | 70 | ||
| GenBank mRNA | AK155875.1 | - | 17 - 18 | 1863 | 70 | |
| BC006873.1 | - | 4 - 5 | 296 | 70 | ||
| BC023957.1 | - | 17 - 18 | 1963 | 5321190 | 70 | |
| BC051101.1 | - | 18 - 19 | 1750 | 5052849 | 70 | |
| CA315096.1 | - | 5 - 6 | 359 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sidt2
Genomic Map