Assay Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289668.1 | NP_001276597.1 | 25 - 26 | 2727 | 65 | |
| NM_172257.4 | NP_758461.1 | 24 - 25 | 2664 | 65 | ||
| GenBank mRNA | AK155875.1 | - | 24 - 25 | 2568 | 65 | |
| BC006873.1 | - | 11 - 12 | 1001 | 65 | ||
| BC023957.1 | - | 24 - 25 | 2668 | 5321190 | 65 | |
| BC051101.1 | - | 25 - 26 | 2455 | 5052849 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sidt2
Genomic Map