Assay Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293689.1 | NP_001280618.1 | 5 - 6 | 1250 | 64 | |
| NM_172271.2 | NP_758475.1 | 5 - 6 | 1250 | 64 | ||
| NR_121629.1 | 4 - 5 | 837 | 64 | |||
| GenBank mRNA | AK032262.1 | - | 4 - 5 | 627 | 64 | |
| AK083807.1 | - | 5 - 6 | 1151 | 64 | ||
| AK085620.1 | - | 5 - 6 | 984 | 64 | ||
| AK163113.1 | - | 5 - 6 | 1255 | 64 | ||
| AY155578.1 | - | 5 - 6 | 1251 | 64 | ||
| BC033610.1 | - | 5 - 6 | 875 | 64 | ||
| BC141148.1 | - | 4 - 5 | 821 | 9055760 | 64 | |
| BC171962.1 | - | 4 - 5 | 821 | 9054248 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a17
Genomic Map