Assay Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172271.2 | NP_758475.1 | 8 - 9 | 1805 | 79 | |
| GenBank mRNA | AY155578.1 | - | 8 - 9 | 1806 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a17
Genomic Map