Assay Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293689.1 | NP_001280618.1 | 9 - 10 | 1994 | 58 | |
| NM_172271.2 | NP_758475.1 | 9 - 10 | 1985 | 58 | ||
| NR_121629.1 | 8 - 9 | 1581 | 58 | |||
| GenBank mRNA | AK032262.1 | - | 8 - 9 | 1371 | 58 | |
| AK083807.1 | - | 9 - 10 | 1895 | 58 | ||
| AK201669.1 | - | 1 - 2 | 137 | 58 | ||
| AY155578.1 | - | 9 - 10 | 1986 | 58 | ||
| BC033610.1 | - | 9 - 10 | 1619 | 58 | ||
| BC141148.1 | - | 8 - 9 | 1565 | 9055760 | 58 | |
| BC171962.1 | - | 8 - 9 | 1565 | 9054248 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a17
Genomic Map