Assay Details
| Gene Symbol: Mark4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172279.1 | NP_758483.1 | 12 - 13 | 1278 | 79 | |
| GenBank mRNA | AK122565.2 | - | 8 - 9 | 924 | 79 | |
| AK146784.1 | - | 13 - 14 | 1613 | 79 | ||
| AY151083.1 | - | 12 - 13 | 1278 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mark4
Genomic Map