Assay Details
| Gene Symbol: Aim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172393.2 | NP_765981.2 | 5 - 6 | 4037 | 103 | |
| GenBank mRNA | AK041082.1 | - | 4 - 5 | 653 | 103 | |
| AK088102.1 | - | 4 - 5 | 709 | 103 | ||
| BC035551.1 | - | 4 - 5 | 1189 | 103 | ||
| BC059272.1 | - | 4 - 5 | 1194 | 103 | ||
More Information
Set Membership: |
|

Aim1
Genomic Map