Assay Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172405.3 | NP_765993.1 | 6 - 7 | 613 | 121 | |
| GenBank mRNA | AK051816.1 | - | 6 - 7 | 614 | 121 | |
| AK167540.1 | - | 6 - 7 | 644 | 121 | ||
| BC029845.1 | - | 6 - 7 | 618 | 4010744 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam175a
Genomic Map