Assay Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172410.2 | NP_765998.1 | 8 - 9 | 750 | 73 | |
| GenBank mRNA | AK010450.1 | - | 7 - 8 | 701 | 73 | |
| AK028864.1 | - | 8 - 9 | 751 | 73 | ||
| AK045380.1 | - | 609 | 73 | |||
| AK129055.1 | - | 7 - 8 | 700 | 73 | ||
| AK167105.1 | - | 7 - 8 | 701 | 73 | ||
| AK169194.1 | - | 5 - 6 | 471 | 73 | ||
| AK169693.1 | - | 7 - 8 | 720 | 73 | ||
| BC023140.1 | - | 7 - 8 | 696 | 3991335 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nup93
Genomic Map