Assay Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172410.2 | NP_765998.1 | 20 - 21 | 2230 | 95 | |
| GenBank mRNA | AK010450.1 | - | 19 - 20 | 2181 | 95 | |
| AK028864.1 | - | 20 - 21 | 2231 | 95 | ||
| AK045380.1 | - | 2089 | 95 | |||
| AK129055.1 | - | 18 - 19 | 2574 | 95 | ||
| AK167105.1 | - | 19 - 20 | 2181 | 95 | ||
| AK169194.1 | - | 17 - 18 | 1951 | 95 | ||
| AK169693.1 | - | 19 - 20 | 2200 | 95 | ||
| BC023140.1 | - | 19 - 20 | 2176 | 3991335 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nup93
Genomic Map