Assay Details
| Gene Symbol: Zc2hc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172414.4 | NP_766002.2 | 2 - 3 | 1529 | 95 | |
| GenBank mRNA | AK032455.1 | - | 2 - 3 | 1433 | 95 | |
| AK033213.1 | - | 2 - 3 | 1424 | 95 | ||
| AK036602.1 | - | 2 - 3 | 1467 | 95 | ||
| BC049940.1 | - | 2 - 3 | 1443 | 6305959 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28603298 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003178 | AK036602.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95775 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53649 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145138 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164846 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003178 | AK036602.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28603303 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07692903 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145138 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164846 | - | - |
| Whole Mouse Genome G4122A | A_51_P266940 | AK036602.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P266940 | NM_172414.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P214956 | AK032455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P214956 | AK033213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P214956 | AK036602.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P214956 | NM_172414.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P348837 | AK032455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P348837 | AK033213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P348837 | AK036602.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P348837 | BC049940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P348837 | NM_172414.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125484 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145138 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164846 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09952526 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28603298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28603320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10015998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09952522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10068936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10015997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09952341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09942040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10024786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09926890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28603285 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10397345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10397344 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10397343 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453602_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443874_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439391_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10397342 | - | - |
| Mouse Genome 430 2.0 Array | 1443874_at | - | - |
| Mouse Genome 430 2.0 Array | 1453602_at | - | - |
| Mouse Genome 430 2.0 Array | 1439391_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453602_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460744_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443874_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439391_at | - | - |
| Mouse Genome 430 2.0 Array | 1460744_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72350_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72350_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200006047.mm.1 | NM_172414.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006047.mm.1 | NM_172414.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006047.mm.1 | TC1200000830.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200006046.mm.1 | NM_172414.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006046.mm.1 | NM_172414.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006046.mm.1 | TC1200000830.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1443874_at | - | - |
| Mouse Expression Set 430 | 1443874_PM_at | - | - |
| Mouse Expression Set 430 | 1453602_at | - | - |
| Mouse Expression Set 430 | 1453602_PM_at | - | - |
| Mouse Expression Set 430 | 1439391_PM_at | - | - |
| Mouse Expression Set 430 | 1439391_at | - | - |
| Murine Genome U74v2 Set | 161102_PM_at | - | - |
| Murine Genome U74v2 Set | 161102_at | - | - |
| Mouse Genome 430A 2.0 Array | 1443874_at | - | - |
| Mouse Genome 430A 2.0 Array | 1443874_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1443874_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1443874_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439391_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439391_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453602_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453602_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 509810 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4560097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4560097 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560097 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711181 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2711181 | - | - |
| mouse-6 v1.1 (Illumina) | scl42960.3_594-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560097 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42960.3_594-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2711181 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2711181 | - | - |

Zc2hc1c
Genomic Map