Assay Details
| Gene Symbol: Mamstr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172418.2 | NP_766006.2 | 3 - 4 | 232 | 63 | |
| GenBank mRNA | AK036109.1 | - | 1 - 2 | 43 | 63 | |
| AK052976.1 | - | 1 - 2 | 98 | 63 | ||
| BY145171.1 | - | 1 - 2 | 35 | 63 | ||
| DQ534901.1 | - | 3 - 4 | 216 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK036109.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of myotube differentiation
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, RNA polymerase II transcription factor binding
protein binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of myotube differentiation
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, RNA polymerase II transcription factor binding
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129999 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154347 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169852 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110895 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154347 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110895 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129999 | - | - |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK017382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK036109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK052976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | NM_172418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122141 | AK012790.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117407 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129273 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154347 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110895 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10553064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553060 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437739_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431836_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10553057 | - | - |
| Mouse Genome 430 2.0 Array | 1431836_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1431836_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437739_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437739_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74490_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74490_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700007510.mm.1 | TC0700000857.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007510.mm.1 | NM_172418.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007510.mm.1 | NM_172418.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007504.mm.1 | TC0700000857.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007504.mm.1 | NM_172418.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007504.mm.1 | NM_172418.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007508.mm.1 | TC0700000857.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007508.mm.1 | NM_172418.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007508.mm.1 | NM_172418.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007506.mm.1 | TC0700000857.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007506.mm.1 | NM_172418.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007506.mm.1 | NM_172418.2 | Refseq |
| Mouse Expression Set 430 | 1437739_a_at | - | - |
| Mouse Expression Set 430 | 1437739_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437739_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431836_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431836_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437739_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 843866 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2470463 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2470463 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002470463 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2622089 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2622089 | - | - |
| mouse-6 v1.1 (Illumina) | scl0074490.1_328-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2470463 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0074490.1_328-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470463 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2622089 | - | - |

Mamstr
Genomic Map