Assay Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020841.1 | - | 2 - 3 | 199 | 71 | |
| AK034183.1 | - | 2 - 3 | 267 | 71 | ||
| AK038638.1 | - | 2 - 3 | 269 | 71 | ||
More Information
Set Membership: |
|

Sphkap
Genomic Map