Assay Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172430.3 | NP_766018.3 | 3 - 4 | 427 | 71 | |
| GenBank mRNA | AK034183.1 | - | 4 - 5 | 470 | 71 | |
| AK038638.1 | - | 4 - 5 | 472 | 71 | ||
| BC069832.1 | - | 3 - 4 | 393 | 30431760 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sphkap
Genomic Map