Assay Details
| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081756.1 | NP_001075225.1 | 8 - 9 | 1034 | 72 | |
| NM_172484.3 | NP_766072.2 | 6 - 7 | 895 | 72 | ||
| NM_176957.3 | NP_795931.2 | 8 - 9 | 1034 | 72 | ||
| GenBank mRNA | AK053243.1 | - | 6 - 7 | 717 | 72 | |
| AY957601.1 | - | 4 - 5 | 454 | 72 | ||
| AY957602.1 | - | 4 - 5 | 454 | 72 | ||
| BC066095.1 | - | 6 - 7 | 895 | 6856184 | 72 | |
| BC079551.1 | - | 6 - 7 | 895 | 6856184 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckap5
Genomic Map