Assay Details
| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176957.3 | NP_795931.2 | 10 - 11 | 1186 | 101 | |
| GenBank mRNA | AK053243.1 | - | 8 - 9 | 869 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckap5
Genomic Map