Assay Details
| Gene Symbol: Araf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009703.2 | NP_033833.1 | 9 - 10 | 1003 | 73 | |
| GenBank mRNA | AK034861.1 | - | 9 - 10 | 998 | 73 | |
| AK085823.1 | - | 9 - 10 | 969 | 73 | ||
| AK135358.1 | - | 9 - 10 | 969 | 73 | ||
| AK163567.1 | - | 9 - 10 | 972 | 73 | ||
| AK169757.1 | - | 9 - 10 | 985 | 73 | ||
| BC004757.1 | - | 9 - 10 | 959 | 3154293 | 73 | |
| D00024.1 | - | 4 - 5 | 365 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Araf
Genomic Map