Assay Details
| Gene Symbol: Srebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011480.3 | NP_035610.1 | 4 - 5 | 887 | 137 | |
| GenBank mRNA | AB017337.1 | - | 3 - 4 | 703 | 137 | |
| AK052628.1 | - | 4 - 5 | 888 | 137 | ||
| AK150052.1 | - | 4 - 5 | 876 | 137 | ||
| AK169607.1 | - | 4 - 5 | 888 | 137 | ||
| AK216773.1 | - | 2 - 3 | 198 | 137 | ||
| BC056922.1 | - | 4 - 5 | 869 | 6824948 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srebf1
Genomic Map