Assay Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB713187.1 | - | 5 - 6 | 488 | 98 | |
| AK088648.1 | - | 4 - 5 | 627 | 98 | ||
| AK153986.1 | - | 4 - 5 | 681 | 98 | ||
More Information
Set Membership: |
|

Diap2
Genomic Map