Assay Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172493.2 | NP_766081.1 | 18 - 19 | 2385 | 89 | |
| GenBank mRNA | AB713187.1 | - | 19 - 20 | 2180 | 89 | |
| AK083254.1 | - | 11 - 12 | 1317 | 89 | ||
| AK088648.1 | - | 18 - 19 | 2319 | 89 | ||
| AK153986.1 | - | 18 - 19 | 2373 | 89 | ||
| AY312280.1 | - | 18 - 19 | 2322 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap2
Genomic Map